LIF379352758 Life Chemicals Inc.

1-(4-Fluoro-phenyl)-4-hydroxy-1H-pyrazole-3-carboxylic acid ethyl ester

  • MDL number MFCD02361225
  • Cas Number : 636568-08-6
  • Molecular Weight : 250.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart