LIF379352761 Life Chemicals Inc.

4,5,6,7-tetrahydro-1,2-benzoxazole-3-carboxylic acid

  • MDL number MFCD03701438
  • Cas Number : 90005-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart