LIF379352762 Life Chemicals Inc.

Pyrimidine-2-carboxylic acid hydrazide

  • MDL number MFCD00508242
  • Cas Number : 87362-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart