LIF379352763 Life Chemicals Inc.

Pyridazine-3-carboxylic acid hydrazide

  • MDL number MFCD10699097
  • Cas Number : 89463-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart