LIF379352764 Life Chemicals Inc.

2-oxo-1,2,3,4-tetrahydroquinoline-6-sulfonyl chloride

  • MDL number MFCD06655638
  • Cas Number : 66657-42-9
  • Molecular Weight : 245.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart