LIF388152228 Life Chemicals Inc.

N-Benzyl-succinamic acid

  • MDL number MFCD00449649
  • Cas Number : 64984-60-7
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart