LIF388152235 Life Chemicals Inc.

5-(pyridin-3-yl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD07377114
  • Cas Number : 893638-39-6
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart