LIF388152236 Life Chemicals Inc.

Toluene-4-sulfonic acid tetrahydro-pyran-4-ylmethyl ester

  • MDL number MFCD09475806
  • Cas Number : 101691-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart