LIF388152238 Life Chemicals Inc.

Acetic acid 2-formyl-quinolin-8-yl ester

  • MDL number MFCD07364750
  • Cas Number : 36456-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart