LIF388152240 Life Chemicals Inc.

2-(morpholin-4-yl)-2-(thiophen-2-yl)ethan-1-amine

  • MDL number MFCD06373942
  • Cas Number : 851176-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart