LIF388152243 Life Chemicals Inc.

(3-Methyl-2-nitro-3H-imidazol-4-yl)-methanol

  • MDL number MFCD06738747
  • Cas Number : 39070-14-9
  • Molecular Weight : 157.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart