LIF388152248 Life Chemicals Inc.

2-(pyrimidin-5-yl)acetic acid

  • MDL number MFCD09608122
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart