LIF388152249 Life Chemicals Inc.

C-(5-Furan-2-yl-1-methyl-1H-pyrazol-3-yl)-methylamine

  • MDL number MFCD10036205
  • Cas Number : 1177358-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart