LIF388152251 Life Chemicals Inc.

Pyrazolo[1,5-a]pyridine-2-carboxylic acid

  • MDL number MFCD06739057
  • Cas Number : 63237-88-7
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart