LIF388152252 Life Chemicals Inc.

N-tert-Butyl-succinamic acid

  • MDL number MFCD00029847
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(tert-butylamino)-4-oxobutanoic acid
Inchi Code 1S/C8H15NO3/c1-8(2,3)9-6(10)4-5-7(11)12/h4-5H2,1-3H3,(H,9,10)(H,11,12)
Inchi Key PDGXYMRZBNNOTC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart