LIF388583758 Life Chemicals Inc.

Methyl 3-aminoadamantane-1-carboxylate hydrochloride

  • MDL number MFCD01826591
  • Cas Number : 80110-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart