LIF388583761 Life Chemicals Inc.

Tetrahydrothiophene-3,4-diamine 1,1-dioxide dihydrochloride

  • MDL number MFCD00456656
  • Cas Number : 70519-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart