LIF388583771 Life Chemicals Inc.

[2-(2,5-Dimethyl-1H-indol-3-yl)ethyl](pyridin-4-ylmethyl)amine dihydrochloride

  • MDL number MFCD04448791
  • Cas Number : 1052409-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart