LIF388583772 Life Chemicals Inc.

4,5,6,7-Tetrahydro-3H-imidazo[4,5-c]pyridine-6-carboxylic acid hydrochloride

  • MDL number MFCD04971929
  • Cas Number : 191327-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart