LIF388583773 Life Chemicals Inc.

1,8-Naphthyridine-2-carboxylic acid hydrobromide

  • MDL number MFCD24387563
  • Cas Number : 1421454-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart