LIF388583774 Life Chemicals Inc.

3-(4-Methylpiperazin-1-yl)propanoic acid dihydrochloride

  • MDL number MFCD08436216
  • Cas Number : 59695-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart