LIF388583784 Life Chemicals Inc.

2-(5-Fluoro-2-methyl-1H-indol-3-yl)-ethylamine hydrochloride

  • MDL number MFCD02615233
  • Cas Number : 1048343-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart