LIF388583786 Life Chemicals Inc.

([1,2,4]triazolo[4,3-a]pyridin-3-ylmethyl)amine hydrochloride

  • MDL number MFCD11052892
  • Cas Number : 1314394-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart