LIF388583788 Life Chemicals Inc.

Adamantane-1,3-diamine dihydrochloride

  • MDL number MFCD00137394
  • Cas Number : 26562-81-2
  • Molecular Weight : 239.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart