LIF388583789 Life Chemicals Inc.

5,6,7,8-Tetrahydro-2H-cinnolin-3-one hydrobromide

  • MDL number MFCD06809755
  • Cas Number : 108749-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart