LIF388583791 Life Chemicals Inc.

2-{[4-(Chloromethyl)-1,3-thiazol-2-yl]methyl}-1,3-benzothiazole hydrochloride

  • MDL number MFCD11519393
  • Cas Number : 1331303-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart