LIF388583793 Life Chemicals Inc.

3-(2-Ethyl-1H-imidazol-1-yl)propanoic acid hydrochloride

  • MDL number MFCD08730446
  • Cas Number : 1171345-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart