LIF388583794 Life Chemicals Inc.

{[5-(1-Benzofuran-2-yl)isoxazol-3-yl]methyl}amine hydrochloride

  • MDL number MFCD11519390
  • Cas Number : 1219211-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart