LIF388583799 Life Chemicals Inc.

3-(aminomethyl)benzenesulfonamide hydrochloride

  • MDL number MFCD10686807
  • Cas Number : 670280-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart