LIF388583802 Life Chemicals Inc.

5-azetidin-3-yl-3-phenyl-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD16652922
  • Cas Number : 1351654-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart