LIF388583804 Life Chemicals Inc.

4-(5-azetidin-3-yl-1,2,4-oxadiazol-3-yl)pyridine dihydrochloride

  • MDL number MFCD16652927
  • Cas Number : 1351602-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart