LIF388583808 Life Chemicals Inc.

4H,5H,6H,7H-thieno[3,2-c]pyridine hydrochloride

  • MDL number MFCD05861485
  • Cas Number : 28783-41-7
  • Molecular Weight : 175.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart