LIF388583809 Life Chemicals Inc.

4-(chloromethyl)-N-phenyl-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD03160812
  • Cas Number : 41981-20-8
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart