LIF388583810 Life Chemicals Inc.

4-(chloromethyl)-N-(3-methoxyphenyl)-1,3-thiazol-2-amine hydrochloride

  • MDL number MFCD22587565
  • Cas Number : 1351602-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart