LIF388583812 Life Chemicals Inc.

1-[(4-methyl-1,3-thiazol-2-yl)methyl]piperazine hydrochloride

  • MDL number MFCD22587589
  • Cas Number : 1351642-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart