LIF388583813 Life Chemicals Inc.

3-(piperidin-4-ylmethyl)imidazolidine-2,4-dione hydrochloride

  • MDL number MFCD22587610
  • Cas Number : 1351647-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart