LIF388583817 Life Chemicals Inc.

4-(1H-1,2,4-triazol-1-ylmethyl)piperidine dihydrochloride

  • MDL number MFCD22587709
  • Cas Number : 1351659-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart