LIF388583819 Life Chemicals Inc.

N-[4-(chloromethyl)-1,3-thiazol-2-yl]pyrimidin-2-amine hydrochloride

  • MDL number MFCD22587711
  • Cas Number : 1351623-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart