LIF388583821 Life Chemicals Inc.

4-amino-1-benzylpyrrolidin-2-one hydrochloride

  • MDL number MFCD09743237
  • Cas Number : 478832-05-2
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart