LIF388583823 Life Chemicals Inc.

5-Aminomethyl-3-phenyl-oxazolidin-2-one hydrochloride

  • MDL number MFCD09743248
  • Cas Number : 1177334-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart