LIF388583827 Life Chemicals Inc.

{[1-(4-Fluorophenyl)-1H-tetrazol-5-yl]methyl}amine hydrochloride

  • MDL number MFCD09743261
  • Cas Number : 1177290-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart