LIF388583834 Life Chemicals Inc.

methyl amino(5-bromo-2-methoxyphenyl)acetate hydrochloride

  • MDL number MFCD16661104
  • Cas Number : 1351581-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart