LIF388583835 Life Chemicals Inc.

1-methyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD07626616
  • Cas Number : 127107-23-7
  • Molecular Weight : 133.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart