LIF388583836 Life Chemicals Inc.

(2-methoxyphenyl)hydrazine hydrochloride

  • MDL number MFCD00035456
  • Cas Number : 6971-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart