LIF388583837 Life Chemicals Inc.

1-(2-morpholin-4-ylethyl)-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD16661008
  • Cas Number : 1351658-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart