LIF388583838 Life Chemicals Inc.

[(4-phenyl-4H-1,2,4-triazol-3-yl)methyl]amine dihydrochloride

  • MDL number MFCD22587834
  • Cas Number : 1351584-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart