LIF388583840 Life Chemicals Inc.

3-[4-(trifluoromethyl)benzyl]pyrrolidine oxalate

  • MDL number MFCD04117741
  • Cas Number : 957998-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart