LIF388583844 Life Chemicals Inc.

1-(3-Pyridylmethyl)piperazine trihydrochloride

  • MDL number MFCD09751277
  • Cas Number : 333992-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart