LIF388583849 Life Chemicals Inc.

N-(3-Aminophenyl)methanesulfonamide hydrochloride

  • MDL number MFCD11519399
  • Cas Number : 57005-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart