LIF388583852 Life Chemicals Inc.

[2-(3,5-dimethyl-1H-pyrazol-1-yl)ethyl]amine hydrobromide

  • MDL number MFCD12401749
  • Cas Number : 663941-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart